ContactUsLinkComponent
Generator, arbitrary functions, 0.1Hz ~ 80MHz with 0.1µHz resolution, AM/FM/FSK, sweep, burst. sine, square, ramp, noise , arbitrary wavefrom signals, 1%~99% adjustable duty cycle, 4.3 inches LCD screen, USB device, RS-232 gpib UK. 1 * 1 items
Catalog # DIDAPMD200050_UK
Supplier:  DIDALAB
Generator, arbitrary functions, 0.1Hz ~ 80MHz with 0.1µHz resolution, AM/FM/FSK, sweep, burst. sine, square, ramp, noise , arbitrary wavefrom signals, 1%~99% adjustable duty cycle, 4.3 inches LCD screen, USB device, RS-232 gpib UK. 1 * 1 items
Catalog # DIDAPMD200050_UK
Supplier:  DIDALAB
Frequently Bought Together