ContactUsLinkComponent
Generator, arbitrary functions, 0.1Hz 12MHz with 0.1Hz resolution, AM/FM/FSK modulation-sweep and frequency counter, sine, square, ramp, noise , arbitrary wavefrom signals, 1%99% adjustable duty cycle, 3.5 inches LCD screen, US 1 * 1 items
  DIDAPMD200020
 :  DIDALAB
Generator, arbitrary functions, 0.1Hz 12MHz with 0.1Hz resolution, AM/FM/FSK modulation-sweep and frequency counter, sine, square, ramp, noise , arbitrary wavefrom signals, 1%99% adjustable duty cycle, 3.5 inches LCD screen, US 1 * 1 items
  DIDAPMD200020
 :  DIDALAB
Frequently Bought Together