ContactUsLinkComponent
Telescope, achromatic lens with 173mm focal length, glareproofed, pulling adjustment, view range from 400mm to infinite, supplied wi th 10x eyepiece with crosshair and 10x micrometer eyepiece, mounted on diameter 10mm rod. 1 * 1 items
Catalog # DIDAPOD069400
Supplier:  DIDALAB
Telescope, achromatic lens with 173mm focal length, glareproofed, pulling adjustment, view range from 400mm to infinite, supplied wi th 10x eyepiece with crosshair and 10x micrometer eyepiece, mounted on diameter 10mm rod. 1 * 1 items
Catalog # DIDAPOD069400
Supplier:  DIDALAB
Frequently Bought Together