ContactUsLinkComponent
Collimator, with lighting, achromatic lens with 230mm focal length, glareproofed, adjustment with knurled ring, lighted crosshair, a uxiliary lighting 6V–2.4W, mounted on diameter 10mm rod. 1 * 1 items
Catalog # DIDAPOD069395
Supplier:  DIDALAB
Collimator, with lighting, achromatic lens with 230mm focal length, glareproofed, adjustment with knurled ring, lighted crosshair, a uxiliary lighting 6V–2.4W, mounted on diameter 10mm rod. 1 * 1 items
Catalog # DIDAPOD069395
Supplier:  DIDALAB
Frequently Bought Together