ContactUsLinkComponent
Gratings transmission, 5 lines/mm, these accuracy gratings have a great uniformity of lines on the whole working area, the working a rea (36x24mm) is protected by a glass plate, compatible with our slide-holders. 1 * 1 items
Catalog # DIDAPOD062400
Supplier:  DIDALAB
Gratings transmission, 5 lines/mm, these accuracy gratings have a great uniformity of lines on the whole working area, the working a rea (36x24mm) is protected by a glass plate, compatible with our slide-holders. 1 * 1 items
Catalog # DIDAPOD062400
Supplier:  DIDALAB
Frequently Bought Together