ContactUsLinkComponent
The spectrogoniometer consist of adjustable focus by knurled screw, achromatic lens, diameter 30mm, with antirefl ective coating, au tocollimating telescope adjustable focus by knurled screw, focus length 162mm, sliding eyepiece. 1 * 1 items
Catalog # DIDAPOD068079
Supplier:  DIDALAB
The spectrogoniometer consist of adjustable focus by knurled screw, achromatic lens, diameter 30mm, with antirefl ective coating, au tocollimating telescope adjustable focus by knurled screw, focus length 162mm, sliding eyepiece. 1 * 1 items
Catalog # DIDAPOD068079
Supplier:  DIDALAB
Frequently Bought Together