ContactUsLinkComponent
Goniometer, used for experiments on prism or grating deviation, 1 minute of arc, adjustable focus by knurled screw, focus length 162 mm, sliding eyepiece, adjustable to infinity by autocollimation with a LED auxiliary light. 1 * 1 items
Catalog # DIDAPOD068049
Supplier:  DIDALAB
Goniometer, used for experiments on prism or grating deviation, 1 minute of arc, adjustable focus by knurled screw, focus length 162 mm, sliding eyepiece, adjustable to infinity by autocollimation with a LED auxiliary light. 1 * 1 items
Catalog # DIDAPOD068049
Supplier:  DIDALAB
Frequently Bought Together