ContactUsLinkComponent
Zinc spectral lamp, bulb, ECO27 screw socket. these spectral lamps have an excellent value for money, they allow the acquisition of rare gases at lower cost. 1 * 1 items
Catalog # DIDAPOF010065
Supplier:  DIDALAB
Zinc spectral lamp, bulb, ECO27 screw socket. these spectral lamps have an excellent value for money, they allow the acquisition of rare gases at lower cost. 1 * 1 items
Catalog # DIDAPOF010065
Supplier:  DIDALAB
Frequently Bought Together