Wardrobe/filing cabinets
Product Details & Documents
Locker/combination cabinet with centre wall. Completely equipped with shelving on the left side, right clothes rail and two additional storage compartments. With clothes hook and door mirror. Carcass construction, i.e. each cabinet can stand on its own, carcass glued.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.
Doors: 2 swing doors with sealing lip, opening angle 125 degrees.
Panels/base: 22 mm thick, with 2 mm ABS edging on the front. Height adjustable in 32 mm increments with recessed shelf supports. Sides and fronts 19 mm thick.
Lock: 3-point locking mechanism with door damper buffer.
Hinges: High quality, all metal cup hinges manufactured by Hettich. Quick mounting technology, 3-dimensionally adjustable.
Delivery information: The cabinets are delivered fully assembled.
Specifications
- Seq:11
- W×D×H:800×420×2156 mm
Complementary Products
Product Information
- Catalog #DescriptionAvailabilityPrice
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



