Orders LinkContactUsLinkComponent
Microplates, 384-well, non binding | Avantor
Microplates, 384-well, non binding
384-well microplate, non binding, flat μClear® bottom, White/clear
Catalog # 738-0176
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 384-well, non binding
Catalog # 738-0176
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 384-well, non binding
384-well microplate, non binding, flat μClear® bottom, White/clear
Price
510,00 €
Log in for availability
1 * 40 ST
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
All plates are free from RNase, DNase, and pyrogens, and are also free of DNA polymerase inhibitors.
Specifications
  • Colour:
    White/clear
  • Description:
    384-well microplate, non binding, flat μClear® bottom
  • Seq:
    05
  • Cat. No.:
    738-0176
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact