Squeegees, Plastic
Plastic squeegee with rubber blade, White, 610 mm
Artikelnr. MICRSQWH-24R
Anbieter: MICRONOVA
Jump to:
Product Details & Documents
These plastic, double foam squeegees are easy to sanitise.
- Will not rust
- Extremely durable
- Synthetic rubber handle with PP frame
- For use with HACCP compliance programs and colour coding
Integrated blade eliminates seams and crevices, reducing places for bacteria to grow. Frame handle is threaded for use with STA series of plastic handles.
Specifications
- Colour:White
- Description:Plastic squeegee with rubber blade
- Length:610 mm
- Seq:05



