ContactUsLinkComponent
Potassium K - 1000 mg/l Standard for AAS in 2% HCl / 12% éthanol 1 * 500 mL
Catalog # ACSDA028.2C12EL.L5
Supplier:  CPACHEM
Potassium K - 1000 mg/l Standard for AAS in 2% HCl / 12% éthanol 1 * 500 mL
Catalog # ACSDA028.2C12EL.L5
Supplier:  CPACHEM