Order Entry
Austria
Orders LinkContactUsLinkComponent
 

ICP multi-element standards (28 components)

ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn, 5% HNO₃, 500 mL
Catalog # ACSDMB56A.K1.5N.L5
Supplier:   CPACHEM
ICP multi-element standards (28 components)
Catalog # ACSDMB56A.K1.5N.L5
Supplier:  CPACHEM
CAS Number:  
ICP multi-element standards (28 components)
ICP multi-element standards (28 components)

ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn, 5% HNO₃, 500 mL

CAS Number:  
Price
799,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

 

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    500 mL
  • Description:
    ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn

Specifications