Order Entry
Austria
Orders LinkContactUsLinkComponent
ICP multi-element standards (28 components)

ICP multi-element standards (28 components)

ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn, 5% HNO₃, 100 mL
Catalog # ACSDMB56A.K1.5N.L1
Supplier:   CPACHEM
ICP multi-element standards (28 components)
Catalog # ACSDMB56A.K1.5N.L1
Supplier:  CPACHEM
CAS Number:  
ICP multi-element standards (28 components)
ICP multi-element standards (28 components)

ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn, 5% HNO₃, 100 mL

CAS Number:  
Price
382,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    ICP multi-element standard, 100 mg/l, (28 elements in in 5 % HNO₃) Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.