Orders LinkContactUsLinkComponent
Roll mop, PolyTack®
Roll mop, PolyTack®
Tacky roller, foam for contoured surfaces, 457,2 mm, 20 sheets/roll
Catalog # CONNPR-18F
Roll mop, PolyTack®
Catalog # CONNPR-18F
Roll mop, PolyTack®
Roll mop, PolyTack®
Tacky roller, foam for contoured surfaces, 457,2 mm, 20 sheets/roll
Price
82,70 €
Log in for availability
1 * 1 Rolle
Product Details & Documents
Tacky roller, with foam or film for cleaning walls, floors, ceiling and hard to reach areas.
  • Easy to use hand or pole attachments
  • Plastic inner core insures cleanliness
  • Anti-microbial agent providing protection against bacterial growth
Specifications
  • Description:
    Tacky roller, foam for contoured surfaces, 457,2 mm
  • Seq:
    03
  • Packed:
    20 sheets/roll