Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Storage Boxes, Multipurpose

94 l, White, 660×450×410 mm
  BURK3418-0194
 :   BURKLE
Storage Boxes, Multipurpose
  BURK3418-0194
 :  BURKLE
Storage Boxes, Multipurpose
Storage Boxes, Multipurpose

94 l, White, 660×450×410 mm

 
101,00 €
   
1 * 1 ST
 

Made of HDPE, these practical storage containers are ideal for safe transport of pourable or liquid contents.

  • With closed gripping edges
  • Can be stacked
  • Rounded inner edges and smooth inner surfaces for easy cleaning
  • Enables use in foodstuffs/hygiene sector
 
  • Capacity:
    94 l
  • Colour:
    White
  • Seq:
    03
  • W×D×H:
    660×450×410 mm
Accessories

 

  •  
    Description
    W×D×H
     
     
  • BURK3418-0001
    Lid for universal storage containers, PS, white
    - mm
       
    1 * 1 ST - 26,50 €