Order Entry
Austria
ContactUsLinkComponent
Calibration gas 25 h2s,100co,2.2%ch4,20.9%o2,n2 34l 4ga20x22w25h100-34 1 * 1 items
  BWTE4GA20X22W25H10
Calibration gas 25 h2s,100co,2.2%ch4,20.9%o2,n2 34l 4ga20x22w25h100-34 1 * 1 items
  BWTE4GA20X22W25H10
Frequently Bought Together