Orders LinkContactUsLinkComponent
Inserts
E 468, insert 1/4 mesh basket with lid and divisions, for various utensils, stainless steel welded mesh, G 7882, G 7892, G 7882 CD, G 7835/36 CD, 250×170×76 mm
Catalog # MIEL5043640
Supplier:   MIELE
Inserts
Catalog # MIEL5043640
Supplier:  MIELE
Inserts
Inserts
E 468, insert 1/4 mesh basket with lid and divisions, for various utensils, stainless steel welded mesh, G 7882, G 7892, G 7882 CD, G 7835/36 CD, 250×170×76 mm
Price
296,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 468, insert 1/4 mesh basket with lid and divisions, for various utensils, stainless steel welded mesh
  • Seq:
    16
  • W×D×H:
    250×170×76 mm
  • For:
    G 7882, G 7892, G 7882 CD, G 7835/36 CD