ContactUsLinkComponent
High performance thermometer (thermistor) with BioCote® antimicrobial protection, selectable C/F scales with default to °C, clock, c ountdown timer, data hold and auto switch off. -50°C to +150°C, -58°F to +302°F thermistor. Lumberg connector. 1 * 1 items
Artikelnr. COMAC21
Lieferant:  COMARK
High performance thermometer (thermistor) with BioCote® antimicrobial protection, selectable C/F scales with default to °C, clock, c ountdown timer, data hold and auto switch off. -50°C to +150°C, -58°F to +302°F thermistor. Lumberg connector. 1 * 1 items
Artikelnr. COMAC21
Lieferant:  COMARK
Frequently Bought Together