Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Storage Boxes for Vials

10×10, Orange, 140×153×55 mm
  NEOL2-1940
 :   NEOLAB
Storage Boxes for Vials
  NEOL2-1940
 :  NEOLAB
Storage Boxes for Vials
Storage Boxes for Vials

10×10, Orange, 140×153×55 mm

 
11,99 €
   
1 * 1 ST
 

Storage boxes, made of PP, coloured, for HPLC vials, cryotubes or 1,5 and 2,0 ml microcentrifuge tubes with the height of 46 mm and up to 12 mm diameter.

  • Stackable
  • Transparent, tight sealing and hinged lid
  • Also usable as cryobox
  • Autoclavable
 
  • Compartments:
    10×10
  • W×D×H ext.:
    140×153×55 mm
  • Colour:
    Orange
  • Seq:
    05