Orders LinkContactUsLinkComponent
Inserts
Inserts
E 363 insert with lid, for upper and lower baskets, for GYN and ENT instruments, G 7831, PG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592, 150×225×55 mm
Catalog # 141-1291
Supplier:   MIELE
Inserts
Catalog # 141-1291
Supplier:  MIELE
Inserts
Inserts
E 363 insert with lid, for upper and lower baskets, for GYN and ENT instruments, G 7831, PG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592, 150×225×55 mm
Price
326,21 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 363 insert with lid, for upper and lower baskets, for GYN and ENT instruments
  • Seq:
    54
  • W×D×H:
    150×225×55 mm
  • For:
    G 7831, PG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592