Orders LinkContactUsLinkComponent
Separating Funnels, Squibb
1000 ml, 29/32 NS
  KAVA632426355946
 :   Kavalier
Separating Funnels, Squibb
  KAVA632426355946
 :  Kavalier
Separating Funnels, Squibb
Separating Funnels, Squibb
1000 ml, 29/32 NS
1 010,00 €
1 * 2 ST
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml
  • Seq:
    05