Orders LinkContactUsLinkComponent
Separating Funnels, Squibb
Separating Funnels, Squibb
50 ml, 19/26 NS
Catalog # KAVA632426355056
Supplier:   Kavalier
Separating Funnels, Squibb
Catalog # KAVA632426355056
Supplier:  Kavalier
Separating Funnels, Squibb
Separating Funnels, Squibb
50 ml, 19/26 NS
Price
243,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    50 ml
  • Seq:
    01