L(+)-Ascorbic acid, Millipore®
Glass bottle for solids, 1 kg
Catalog # 1831-1KG
Supplier: Merck Millipore (Calbiochem)
L(+)-Ascorbic acid, Millipore®
Catalog # 1831-1KG
Supplier: Merck Millipore (Calbiochem)
CAS Number: 50-81-7
Springe zu:
Product Details & Documents
Synonyme: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Nummer:50-81-7
- Formel:C₆H₈O₆
- MDL-Nummer:MFCD00064328
- Molekulargewicht:176.13 g/mol
- Merck-Index:13,00837
- Siedepunkt:553 °C (1013 hPa)
- Schmelzpunkt:190…192 °C
- Dichte:1.694 g/cm³ (20 °C)
- Lagertemperatur:Ambient
- EINECS:200-066-2
- REACH:exempt
Spezifikationen
Spezifikationen
- Pack type:Glass bottle for solids
- Pk:1 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



