Orders LinkContactUsLinkComponent
Peqlab, PCR Tubes | Avantor
Peqlab, PCR Tubes
PCR tubes with domed cap, Clear, 0,2 ml, 2 bags of 500
Catalog # 732-3206
   Low Manufacturing Impact
Supplier:   VWR Collection
Peqlab, PCR Tubes
Catalog # 732-3206
Supplier:  VWR Collection
   Low Manufacturing Impact
Peqlab, PCR Tubes
Peqlab, PCR Tubes
PCR tubes with domed cap, Clear, 0,2 ml, 2 bags of 500
Price
46,60 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Ultra-thin walls for rapid heat transfer
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.
Specifications
  • Packed:
    2 bags of 500
  • Capacity:
    0,2 ml
  • Colour:
    Clear
  • Description:
    PCR tubes with domed cap
  • Cat. No.:
    732-3206
  • Environmental certification:
    IRECS
  • Environmentally Preferable:
    Low Manufacturing Impact
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.