Order Entry
Austria
Orders LinkContactUsLinkComponent
Blocker™ Blocking Buffer, Casein

Blocker™ Blocking Buffer, Casein

Blocker™ Casein in TBS, blocking buffer, 100 mL
Catalog # PIER37583
Blocker™ Blocking Buffer, Casein
Catalog # PIER37583
CAS Number:  
Blocker™ Blocking Buffer, Casein
Blocker™ Blocking Buffer, Casein

Blocker™ Casein in TBS, blocking buffer, 100 mL

CAS Number:  
Price
57,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Blocker™ Casein in PBS or TBS are ready-to-use, solutions of purified casein protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.

  • Easy to use
  • Safe, stable and thimerosal-free formulations
  • Purified casein provides fewer chances of cross reaction
  • Flexible and available in PBS and TBS formulations

This blocking buffer contains casein protein that is purified from milk by the Hammarsten method. The phosphate and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody based detection methods. Blocker Casein buffers are 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Caution: For research use only. Not for use in diagnostic procedures.

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Blocker™ Casein in TBS, blocking buffer
  • Cat. No.:
    PIER37583
  • Physical form:
    Liquid
  • Concentration:
    1X
  • Storage buffer:
    TBS
  • For use with:
    Western blot, ELISA

Specifications