Order Entry
Austria
Orders LinkContactUsLinkComponent
SuperBlock™ Blocking Buffer

SuperBlock™ Blocking Buffer

SuperBlock® (PBS) blocking buffer, 100 mL
Catalog # PIER37580
SuperBlock™ Blocking Buffer
Catalog # PIER37580
CAS Number:  
SuperBlock™ Blocking Buffer
SuperBlock™ Blocking Buffer

SuperBlock® (PBS) blocking buffer, 100 mL

CAS Number:  
Price
64,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

SuperBlock Blocking buffer is an optimised solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.

  • Features of SuperBlock Blocking Buffer include:
  • Blocking time - block membranes in five to ten minutes and ELISA plates in two minutes
  • Application - ELISA, western blotting, IHC biotin-free for use with streptavidin system
  • Stability - store buffer at 4 ℃ for one year store blocked plates dry for up to 12 months

Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilising them for drying and storage for later use.

Caution: For research use only. Not for use in diagnostic procedures.

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    SuperBlock® (PBS) blocking buffer
  • Cat. No.:
    PIER37580
  • Physical form:
    Liquid
  • Concentration:
    1X
  • Storage buffer:
    PBS
  • For use with:
    ELISA, Western blot

Specifications