Order Entry
Austria
Orders LinkContactUsLinkComponent
Blocker™ Blocking Buffer, Casein

Blocker™ Blocking Buffer, Casein

Blocker™ Casein in PBS, blocking buffer, 100 mL
Catalog # PIER37582
Blocker™ Blocking Buffer, Casein
Catalog # PIER37582
CAS Number:  
Blocker™ Blocking Buffer, Casein
Blocker™ Blocking Buffer, Casein

Blocker™ Casein in PBS, blocking buffer, 100 mL

CAS Number:  
Price
57,50 €
Log in for availability
1 * 100 mL
Product Details & Documents

Blocker™ Casein in PBS or TBS are ready-to-use, solutions of purified casein protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.

  • Easy to use
  • Safe, stable and thimerosal-free formulations
  • Purified casein provides fewer chances of cross reaction
  • Flexible and available in PBS and TBS formulations

This blocking buffer contains casein protein that is purified from milk by the Hammarsten method. The phosphate and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody based detection methods. Blocker Casein buffers are 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Caution: For research use only. Not for use in diagnostic procedures.

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Blocker™ Casein in PBS, blocking buffer
  • Cat. No.:
    PIER37582
  • Physical form:
    Liquid
  • Concentration:
    1X
  • Storage buffer:
    PBS
  • For use with:
    Western blot, ELISA

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.