The helium-specific triple trap is ideal for purifying helium in gc/ms systems. this trap is packed and purged under helium and cont ains oxygen, moisture and hydrocarbon scrubbers in one cartridge. its features and benefits include - ultra-high capa 1 * 1 items
Catalog # PERCN9306820
Supplier: PerkinElmer
Frequently Bought Together