DIMETHYLDIOCTYLAMMONIUM CHLORIDE 1 * 0,1 g
Catalog # EHERC12726470
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Water Reag. Ph. Eur. 1095502 carbon dioxide-free
Supplier: VWR Chemicals
VWR®, Viscosity standards
Supplier: VWR Chemicals
Colour Reference Solutions, Reag.Ph.Eur.
Supplier: REAGECON
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
Xylenol orange triturate 1% (w/w) in potassium nitrate Reag. Ph. Eur. 1096301
Supplier: VWR Chemicals
AMOXICILLIN TRIHYDRATE PERFORMANCE VERIF
Supplier: EDQM
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
DL-α-Tocopherol acetate
Supplier: EDQM