Order Entry
Austria
Orders LinkContactUsLinkComponent
Process documentaion

Process documentaion

APH 302 Connection cable type 2 for connection of a machine with serial interface to a PC. Null modem cable, length 3 m, Ø 4×185 mm, All glassware washers
Catalog # 141-0617
Supplier:   MIELE
Process documentaion
Catalog # 141-0617
Supplier:  MIELE
Process documentaion
Process documentaion

APH 302 Connection cable type 2 for connection of a machine with serial interface to a PC. Null modem cable, length 3 m, Ø 4×185 mm, All glassware washers

Price
33,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    APH 302 Connection cable type 2 for connection of a machine with serial interface to a PC. Null modem cable, length 3 m, Ø 4×185 mm
  • Seq:
    13
  • For:
    All glassware washers
  • Cat. No.:
    141-0617
  • W×D×H:
    Ø 4×185 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.