Order Entry
Austria
Orders LinkContactUsLinkComponent
Process documentaion

Process documentaion

APH 303 Extension cable 3 m, for the extension of connection cable type 1 and 2, Ø 2,5×125 mm, All glassware washers
Catalog # 141-0619
Supplier:   MIELE
Process documentaion
Catalog # 141-0619
Supplier:  MIELE
Process documentaion
Process documentaion

APH 303 Extension cable 3 m, for the extension of connection cable type 1 and 2, Ø 2,5×125 mm, All glassware washers

Price
32,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    APH 303 Extension cable 3 m, for the extension of connection cable type 1 and 2, Ø 2,5×125 mm
  • Seq:
    15
  • For:
    All glassware washers
  • Cat. No.:
    141-0619
  • W×D×H:
    Ø 2.5×125 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.