Order Entry
Austria
Orders LinkContactUsLinkComponent
Process documentaion

Process documentaion

APH 406 Network cable for connection to the network, length 3 m (type 6), Transparent silver, G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
  141-0623
 :   MIELE
Process documentaion
  141-0623
 :  MIELE
Process documentaion
Process documentaion

APH 406 Network cable for connection to the network, length 3 m (type 6), Transparent silver, G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593

 
39,00 €
   
1 * 1 ST
 
 
  • Description:
    APH 406 Network cable for connection to the network, length 3 m (type 6), Transparent silver
  • Seq:
    18
  • For:
    G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
  • Cat. No.:
    141-0623
  • W×D×H:
    195×479×72 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.