Order Entry
Austria
Orders LinkContactUsLinkComponent
Process documentaion

Process documentaion

APH 408 Crossover network cable for direct connection to PC, length 3 m (type 8), G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
Catalog # 141-0625
Supplier:   MIELE
Process documentaion
Catalog # 141-0625
Supplier:  MIELE
Process documentaion
Process documentaion

APH 408 Crossover network cable for direct connection to PC, length 3 m (type 8), G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593

Price
39,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    APH 408 Crossover network cable for direct connection to PC, length 3 m (type 8)
  • Seq:
    20
  • For:
    G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
  • Cat. No.:
    141-0625
  • W×D×H:
    200×350×80 mm