Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Process documentaion

APH 304 Extension cable 5 m, for the extension of connection cable type 1 and 2, All glassware washers
  141-0620
 :   MIELE
Process documentaion
  141-0620
 :  MIELE
Process documentaion
Process documentaion

APH 304 Extension cable 5 m, for the extension of connection cable type 1 and 2, All glassware washers

 
33,00 €
   
1 * 1 ST
 
 
  • Description:
    APH 304 Extension cable 5 m, for the extension of connection cable type 1 and 2
  • Seq:
    16
  • For:
    All glassware washers
  • Cat. No.:
    141-0620
  • W×D×H:
    133×479×72 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.