Orders LinkContactUsLinkComponent
Inserts
E 149 Insert suitable for 80 test tubes up to 16×105 mm, 80 Compartments at 18×18 mm and additionally equipped with a lid, W×D×H: 528×527×206 mm
Catalog # 141-0356
Supplier:   MIELE
Inserts
Catalog # 141-0356
Supplier:  MIELE
Inserts
Inserts
E 149 Insert suitable for 80 test tubes up to 16×105 mm, 80 Compartments at 18×18 mm and additionally equipped with a lid, W×D×H: 528×527×206 mm
Price
536,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 149 Insert suitable for 80 test tubes up to 16×105 mm, 80 Compartments at 18×18 mm and additionally equipped with a lid, W×D×H: 528×527×206 mm
  • Seq:
    44