Orders LinkContactUsLinkComponent
Inserts
E 109 Insert suitable for 21 glass beakers up to 250 ml, equipped with 21×3 stay bars
Catalog # 141-0297
Supplier:   MIELE
Inserts
Catalog # 141-0297
Supplier:  MIELE
Inserts
Inserts
E 109 Insert suitable for 21 glass beakers up to 250 ml, equipped with 21×3 stay bars
Price
296,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 109 Insert suitable for 21 glass beakers up to 250 ml, equipped with 21×3 stay bars
  • Seq:
    35