Orders LinkContactUsLinkComponent
Inserts
Inserts
E 111 Insert suitable for 8 glass beakers between 600 - 1000 ml, equipped with 8×3 stay bars
Catalog # 141-0298
Supplier:   MIELE
Inserts
Catalog # 141-0298
Supplier:  MIELE
Inserts
Inserts
E 111 Insert suitable for 8 glass beakers between 600 - 1000 ml, equipped with 8×3 stay bars
Price
217,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 111 Insert suitable for 8 glass beakers between 600 - 1000 ml, equipped with 8×3 stay bars
  • Seq:
    37