Orders LinkContactUsLinkComponent
Inserts
Inserts
E 137 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm
Catalog # 141-0381
Supplier:   MIELE
Inserts
Catalog # 141-0381
Supplier:  MIELE
Inserts
Inserts
E 137 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm
Price
482,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 137 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm
  • Seq:
    50