CoolCell Filler Vials maintain freezing consistency when freezing less than a full load. Insert a CoolCell Filler Vial into each empty well in the CoolCell Freezing container to ensure a consistent 1C/minute freeze rate for all vials. Vials contai 1 * 6 items
Catalog # BCISBCS-3105
Supplier: BIOCISION
Frequently Bought Together