8-Bromoadenine
Glass bottle for solids, 500 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 8-Bromo-1H-adenine, 8-Bromo-7H-purin-6-amine, 8-Bromo-9H-purin-6-amine
- CAS Number:6974-78-3
- Formula:C₅H₄BrN₅
- MDL Number:MFCD00082518
- Molecular Weight:214.02 g/mol
- Melting Pt:>250 °C
- Storage Temperature:Refrigerator
- EINECS:230-225-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



