Order Entry
Austria
Orders LinkContactUsLinkComponent
 

8-Bromoadenine

Glass bottle for solids, 100 mg
Catalog # J65582.MC
Irritant
8-Bromoadenine
Catalog # J65582.MC
CAS Number:  6974-78-3
8-Bromoadenine
8-Bromoadenine

Glass bottle for solids, 100 mg

CAS Number:  6974-78-3
Price
79,70 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  8-Bromo-1H-adenine, 8-Bromo-7H-purin-6-amine, 8-Bromo-9H-purin-6-amine
  • CAS Number:6974-78-3
  • Formula:C₅H₄BrN₅
  • MDL Number:MFCD00082518
  • Molecular Weight:214.02 g/mol
  • Melting Pt:>250 °C
  • Storage Temperature:Refrigerator
  • EINECS:230-225-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications