Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solution, 4,00, ±0,01, 25 °C, Plastic bottle, 500 mL
  REGECC1040525
 :   REAGECON
Buffer solutions
  REGECC1040525
 :  REAGECON
 :  
Buffer solutions
Buffer solutions

Buffer solution, 4,00, ±0,01, 25 °C, Plastic bottle, 500 mL

 :  
 
32,90 €
   
1 * 500 mL
 

Documents

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    4,00

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.