Proxima 2700 Imaging System Specifications: bit depth: 16bit CCD cooling: yes, triple peltier and thermostatic Exposure time: 0.001 - 3600s Resolution: 1.45 M pixels Electronic contol (iris, focus, zoom): yes Zoom: no Image positioning system: no Aut 1 * 1 ST
Catalog # IBSAIM-520-2700
Supplier: IBS Stock Items
Frequently Bought Together