Conductivity cells, IDS
IDS conductivity cell, TetraCon® 925-P, 4-cell graphite, wireless ready, TetraCon® 925-P, 4-cell graphite, wireless ready, 10 µS/cm - 2000 mS/cm, 0,475 cm-1, 0...+100 °C, Epoxy, 15,3×120 mm, Plug head for AS/IDS-x cable or wireless module
665-0304
: WTW
High quality, enhanced sensor technology combined with most up to date measurement electronics. For use with IDS meters.
- Intelligent: Sensors are automatically recognised by the meter, each sensor is individually identified
- Digital: Measurement signals are processed digitally, providing very accurate data
- IDS cells store their serial number and calibration records
- TetraCon® 925: 4-electrode measuring cell ideal for medium and high conductivity applications
- LR 925/01: Ideal for pure water and low conductivity applications
- TetraCon® 925/LV: Graphite 4-electrode cell for small volumes and viscous samples
The electrodes can provide the following data, shown on the screen of the digital meter:
- Model name
- Serial number
- Calibration status (+++, ++, +, -, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values, etc.
: Various cable lengths available.
- Conductivity:10 µS/cm - 2000 mS/cm
- Connection:Plug head for AS/IDS-x cable or wireless module
- Ø×L:15,3×120 mm
- Type:TetraCon® 925-P, 4-cell graphite, wireless ready
- Description:IDS conductivity cell, TetraCon® 925-P, 4-cell graphite, wireless ready
- Material:Epoxy
- Seq:03
- Temp.:0...+100 °C
- Cell constant:0,475 cm-1



