ContactUsLinkComponent
Safety cabinet with wing doors for the storage of pressurised gas cylinders. Outer dimensions: 595 x 596 x 2070 (WxDxH) mm. Inner di mensions: 500 x 404 x 1884 (WxDxH) mm, Weight: approx. 300 kg Wing door hung right Outer carcass in sheet steel powde 1 * 1 items
  DUEH73-200660-411
 :  DUPERTHAL
Safety cabinet with wing doors for the storage of pressurised gas cylinders. Outer dimensions: 595 x 596 x 2070 (WxDxH) mm. Inner di mensions: 500 x 404 x 1884 (WxDxH) mm, Weight: approx. 300 kg Wing door hung right Outer carcass in sheet steel powde 1 * 1 items
  DUEH73-200660-411
 :  DUPERTHAL
Frequently Bought Together