ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.3973g/ml @ 50C 1 * 100 ml
Catalog # REGEDEN50200
Supplier:  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.3973g/ml @ 50C 1 * 100 ml
Catalog # REGEDEN50200
Supplier:  REAGECON
Frequently Bought Together