Sucrose (Brix) Standard 60% - 1.441928 (± 0.00004), Stabilised, shelf-life of 1 year, 6 x 15ml 1 * 90 mL
Catalog # REGEBS60S6
Supplier: REAGECON
Sucrose (Brix) Standard 60% - 1.441928 (± 0.00004), Stabilised, shelf-life of 1 year, 6 x 15ml 1 * 90 mL
Catalog # REGEBS60S6
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
HPLC columns, LiChrospher®
Supplier: Merck
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Capillary columns, HP-FFAP
Supplier: Agilent