Sucrose (Brix) Standard 50% - 1.420087 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
REGEBS50S
: REAGECON
Sucrose (Brix) Standard 50% - 1.420087 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
REGEBS50S
: REAGECON
Frequently Bought Together
pH buffers, ready to use
: WTW
Sulfalene 1 * 100 mg
: EHRENSTORFER
Hesperidin
: EHRENSTORFER
2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123)
: EHRENSTORFER