Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sucrose (Brix) Standard 40% - 1.39986 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS40S
Supplier:
REAGECON
Expand image
Sucrose (Brix) Standard 40% - 1.39986 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS40S
Supplier:
REAGECON
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Dehydrated culture media
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Sucrose (Brix) Standard 0% - 1.332986 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Supplier: REAGECON